missing translation for 'onlineSavingsMsg'
Learn More

Proteintech Viability/Cytotoxicity Assay Kit for Animal Live & Dead Cells (Calcein AM, PI Method)

Viability/Cytotoxicity Assay Kit for Animal Live Dead Cells (Calcein AM, PI method) provides dual fluorescence staining for the detection of living and dead cells.

105.00 € - 199.00 €

Specifications

For Use With (Application) Flow Cytometry
Emission 617 nm, 517 nm
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17551176
View Documents Promotion Details
Proteintech
PF00007-150T
150 Tests
105.00 €
150 tests
Please to purchase this item. Need a web account? Register with us today!
17561176
View Documents Promotion Details
Proteintech
PF00007-300T
300 Tests
199.00 €
300 tests
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The two probes in the kit effect cell viability by measuring intracellular esterase activity and plasma membrane integrity.
  • This kit can be used for fluorescence microscopes, flow cytometers, microplate readers and other fluorescence detection systems.
  • This kit can be used for most eukaryotic mammalian samples, but not to fungi and yeast.
Specifications

Specifications

Flow Cytometry
617 nm, 517 nm
Videos
SDS
Documents

Documents

Product Certifications