Learn More
Scientific Industries SI™ Vertical Microtube Holder
Requires no additional attachment for use. Scientific Industries SI™ Vertical Microtube Holder snaps microtubes into place.
124.00 € - 133.00 €
Specifications
For Use With (Equipment) Vortex-Genie 2 Mixer and Vortex-Genie Pulse
View More Specs
Products 2
Product Code Brand No. of Positions Price Quantity & Availability  
Product Code Brand No. of Positions Price Quantity & Availability  
15587365
View Documents
Scientific Industries SI™
SI-V209
9
124.00 €
Each
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
15517375
View Documents
Scientific Industries SI™
SI-V524
24
133.00 €
Each
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
Description
Description
  • Holds tubes vertically
  • Snaps on to any Vortex-Genie 2 Mixer and the Vortex-Genie Pulse
  • Optimized for high speed vortexing a small number of tubes
  • No additional attachment necessary
Specifications
Specifications
Vortex-Genie 2 Mixer and Vortex-Genie Pulse
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Scientific Industries SI™ Vertical Microtube Holder