missing translation for 'onlineSavingsMsg'
Learn More

Systec™ Vertical Floor-Standing, Top-Loading Autoclave, VB-Series

Systec™ Vertical Floor-Standing, Top-Loading Autoclave, VB-Series is suitable for basic sterilization applications and can accept limited additional process optimization options.

Specifications

Product Type Autoclave, Top-loading Vertical
Temperature (Metric) 140°C
Operating Pressure 4 bar
View More Specs

Description

Description

  • Standard features:
    • Internal heating elements within the autoclave chamber
    • Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
    • LCD display and fully automatic microprocessor control
    • Number of sterilization programs: 4
    • Code-secured access rights for changing parameters and further safety-relevant intervention
    • Flexible PT-100 temperature sensor
  • Optional integrated printer for batch documentation
Specifications

Specifications

Autoclave, Top-loading Vertical
140°C
4 bar
Videos
SDS
Documents

Documents

Product Certifications