Learn More
Ansell™ VersaTouch™ 92-220
Improved manufacturing processes for longer wear, greater tear strength and better grip in dry, wet and fatty conditions. A thinner glove with enhanced fingertip flexibility and improved overall comfort.
19.65 € - 26.70 €
Specifications
Exterior Finish Fully Textured
Material Nitrile
Series 92-220
View More Specs
Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12677061
View Documents
Ansell™
92220100
10 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12647061
View Documents
Ansell™
92220070
7 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12657061
View Documents
Ansell™
92220080
8 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12667061
View Documents
Ansell™
92220090
9 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Improved manufacturing processes for longer wear, greater tear strength and better grip in dry, wet and fatty conditions.
  • A thinner glove with enhanced fingertip flexibility and improved overall comfort.
  • Textured fingertips provide good comfort and tactility for sensitive, precise handling.
  • Latex-free: no risk of Type 1 allergies.
  • Approved for food contact applications. Ideal for handling all fatty food.
Specifications
Specifications
Fully Textured
Nitrile
92-220