Learn More
Veratraldehyde, 99%, Thermo Scientific Chemicals
34.37 € - 366.83 €
Chemical Identifiers
CAS | 120-14-9 |
---|---|
Molecular Formula | C9H10O3 |
Molecular Weight (g/mol) | 166.18 |
MDL Number | MFCD00003363 |
InChI Key | WJUFSDZVCOTFON-UHFFFAOYSA-N |
Synonym | veratraldehyde, veratric aldehyde, methylvanillin, vanillin methyl ether, veratral, veratryl aldehyde, benzaldehyde, 3,4-dimethoxy, veratrum aldehyde, p-veratric aldehyde, 4-o-methylvanillin |
PubChem CID | 8419 |
ChEBI | CHEBI:17098 |
IUPAC Name | 3,4-dimethoxybenzaldehyde |
SMILES | COC1=CC=C(C=O)C=C1OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11418993
|
Thermo Scientific Alfa Aesar
A11110.22 |
100 g |
34.37 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11428993
|
Thermo Scientific Alfa Aesar
A11110.36 |
500 g |
86.53 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11438993
|
Thermo Scientific Alfa Aesar
A11110.0E |
2500 g |
366.83 €
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Veratraldehyde was used in the preparation of 4-chloromethyl-2-(dimethoxyphenyl)-1,3-dioxolane. It was used in the synthesis of (+)-lithospermic acid, having anti-HIV activity. It forms 1:1 inclusion complexes with cyclodextrins. It reacts with 3-acetyl-2,5-dimethythiophene to yield chalcone dye, (2E)-3-(3,4-Dimethoxyphenyl)-1-(2,5-dimethylthiophen-3-yl)prop-2-en-1-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsVeratraldehyde was used in the preparation of 4-chloromethyl-2-(dimethoxyphenyl)-1,3-dioxolane. It was used in the synthesis of (+)-lithospermic acid, having anti-HIV activity. It forms 1:1 inclusion complexes with cyclodextrins. It reacts with 3-acetyl-2,5-dimethythiophene to yield chalcone dye, (2E)-3-(3,4-Dimethoxyphenyl)-1-(2,5-dimethylthiophen-3-yl)prop-2-en-1-one.
Solubility
Soluble in water. Freely soluble in alcohol and ether.
Notes
Air sensitive. Incompatible with oxidizing agents.
Chemical Identifiers
120-14-9 | |
166.18 | |
WJUFSDZVCOTFON-UHFFFAOYSA-N | |
8419 | |
3,4-dimethoxybenzaldehyde |
C9H10O3 | |
MFCD00003363 | |
veratraldehyde, veratric aldehyde, methylvanillin, vanillin methyl ether, veratral, veratryl aldehyde, benzaldehyde, 3,4-dimethoxy, veratrum aldehyde, p-veratric aldehyde, 4-o-methylvanillin | |
CHEBI:17098 | |
COC1=CC=C(C=O)C=C1OC |
Specifications
120-14-9 | |
281°C | |
C9H10O3 | |
473899 | |
14,9951 | |
Soluble in water. Freely soluble in alcohol and ether. | |
COC1=CC=C(C=O)C=C1OC | |
166.18 | |
CHEBI:17098 | |
99% |
42°C to 46°C | |
112°C (233°F) | |
MFCD00003363 | |
Air Sensitive | |
veratraldehyde, veratric aldehyde, methylvanillin, vanillin methyl ether, veratral, veratryl aldehyde, benzaldehyde, 3,4-dimethoxy, veratrum aldehyde, p-veratric aldehyde, 4-o-methylvanillin | |
WJUFSDZVCOTFON-UHFFFAOYSA-N | |
3,4-dimethoxybenzaldehyde | |
8419 | |
166.18 | |
Veratraldehyde |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 204-373-2
missing translation for 'rtecsNumber' : YX5088000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.