Learn More
VACUUBRAND™ VARIO™ PC 3010 NT Select Chemistry Pumping Unit
GREENER_CHOICE
The PC 3010 NT pumping unit precisely controls vacuum level to achieve unparalleled process control. With its combination of high pumping speed and fractional-millibar ultimate vacuum, this pump is suitable for demanding applications, even with high boiling point solvents at low temperatures.
Brand:  VACUUBRAND™ 25744851
 View more versions of this product
Product Code. 17754919
  • 13995.00 € / Each
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • VACUU·SELECT vacuum controller with graphical user interface and predefined applications simplifies work in laboratory
  • Automatic boiling point detection and adjustment of vacuum for short process times
  • Ideal for evaporation of large amounts of high-boiling solvents, even at low temperatures
  • Extra long diaphragm life for minimum operational and servicing costs
  • Environmental compatibility through low energy consumption and efficient solvent recovery
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.6 mbar
47 dBA
29.7 kg
450 mm
387 mm
IP40
1.1 bar
200/230 V
Mounted Pump, Manual
CH
12.8 m3/hr.
616 mm
Vacuum System
Work in Miniplants, Kilo Labs, Scale-up Facilities
Rotary Evaporators, Vacuum Drying Ovens, Vacuum Distillation Columns