missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ VapLock™ Waste Funnels
VapLock™ Waste Funnels safely and securely capture waste and hazardous solvents
Specifications
Type | Waste Funnel, 4 in. dia |
---|---|
For Use With (Application) | Solvent Safety |
Product Code | Brand | For Use With (Equipment) | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Material | Price | Quantity & Availability | ||||
15814534
|
Cole-Parmer™
SF-003 |
Justrite Centura Containers | PP and EPDM quick disconnect, HDPE funnel |
N/A
|
N/A | ||||
15824534
|
Cole-Parmer™
SF-005 |
Justrite Centura Containers | SS and PTFE quick disconnect, HDPE funnel |
N/A
|
N/A | ||||
15834534
|
Cole-Parmer™
SF-010 |
Container Ports with 1/2 in. NPT(F) | HDPE, PP |
N/A
|
N/A | ||||
Description
- Reduces fumes and harmful vapor exhaust
- Easily connect to Justrite™ Centura safety containers
Warranty
None
Specifications
Waste Funnel, 4 in. dia | |
Solvent Safety |