missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Jars and Pails
VapLock™ Solvent Waste Manifold Caps for Jars and Pails connect to waste containers for a secure closed system
Specifications
Type | Solvent Waste Manifold |
---|---|
For Use With (Application) | Solvent Safety |
For Use With (Equipment) | Solvent-Compatible Containers |
Description
- Select from 70 mm Rieke or 100 to 415 cap sizes to suit your container
- Different materials, numbers and sizes of ports help customize to your application
Warranty
None
Specifications
Solvent Waste Manifold | |
Solvent Safety | |
Solvent-Compatible Containers |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title