missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ VapLock™ Solvent Waste Kits for Drums and Panel Mounting

VapLock™ Solvent Waste Kits for Drums and Panel Mounting collect effluent from multiple sources while minimizing harmful solvent exhaust

Specifications

Type Solvent Waste Kit
For Use With (Application) Solvent Safety
For Use With (Equipment) Solvent-Compatible Containers
Thread Size (English) 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F)
View More Specs

Description

Description

Various cap sizes available to fit your solvent container

Warranty

None

Specifications

Specifications

Solvent Waste Kit
Solvent Safety
Solvent-Compatible Containers
1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F)
Videos
SDS
Documents

Documents

Product Certifications