missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ VapLock™ Solvent Delivery Caps with Inlet Valve and Filter
VapLock™ Solvent Delivery Caps with Inlet Valve and Filter reduce chemical evaporation and exposure to hazardous vapors
Specifications
For Use With (Application) | Solvent Safety |
---|---|
For Use With (Equipment) | Solvent-Compatible Containers |
Thread Size (English) | 1/4 in.-28 UNF(F) |
Description
Use with semi-rigid tubing equal to or less than 1/8 in (3.2 mm) ODWarranty
None
Specifications
Solvent Safety | |
Solvent-Compatible Containers | |
1/4 in.-28 UNF(F) |