Learn More
Vanadium(V) trifluoride oxide, 99%, Thermo Scientific™
68.60 € - 236.00 €
Chemical Identifiers
CAS | 13709-31-4 |
---|---|
Molecular Formula | F3H3OV |
Molecular Weight (g/mol) | 126.96 |
MDL Number | MFCD00042555 |
InChI Key | HHQFLEDKAVLHOM-UHFFFAOYSA-N |
Synonym | trifluorooxovanadium, trifluoro oxo vanadium, vanadium oxytrifluoride, vof3, vanadium v trifluoride oxide, vanadium v oxyfluoride, vanadium,trifluorooxo-, t-4-9ciShow More |
PubChem CID | 123322 |
IUPAC Name | oxovanadium;trihydrofluoride |
SMILES | F.F.F.O=[V] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13709-31-4 | |
126.96 | |
HHQFLEDKAVLHOM-UHFFFAOYSA-N | |
123322 | |
F.F.F.O=[V] |
F3H3OV | |
MFCD00042555 | |
trifluorooxovanadium, trifluoro oxo vanadium, vanadium oxytrifluoride, vof3, vanadium v trifluoride oxide, vanadium v oxyfluoride, vanadium,trifluorooxo-, t-4-9ciShow More | |
oxovanadium;trihydrofluoride |
Specifications
300°C | |
Powder | |
MFCD00042555 | |
trifluorooxovanadium, trifluoro oxo vanadium, vanadium oxytrifluoride, vof3, vanadium v trifluoride oxide, vanadium v oxyfluoride, vanadium,trifluorooxo-, t-4-9ci | |
HHQFLEDKAVLHOM-UHFFFAOYSA-N | |
oxovanadium;trihydrofluoride | |
123322 | |
99% | |
Vanadium(V) trifluoride oxide |
13709-31-4 | |
F3H3OV | |
UN3290 | |
Insoluble in water. | |
F.F.F.O=[V] | |
126.96 | |
123.94 | |
Moisture sensitive |
Safety and Handling
GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 237-250-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.