missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV,V) oxide, 99.5% (metals basis), Thermo Scientific™

72.50 € - 262.00 €

Chemical Identifiers

CAS 12037-42-2
Molecular Formula O13V6
Molecular Weight (g/mol) 513.64
MDL Number MFCD00064848
InChI Key FTGZPVKRMCXHDZ-UHFFFAOYSA-N
Synonym vanadium oxide v6o13, vanox 13
PubChem CID 72720435
IUPAC Name dioxovanadiooxy(dioxo)vanadium;dioxovanadium
SMILES O=[V]=O.O=[V]=O.O=[V]=O.O=[V]=O.O=[V](=O)O[V](=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11378227
View Documents
Thermo Scientific Alfa Aesar
022956.09
10 g
72.50 €
10g
Please to purchase this item. Need a web account? Register with us today!
11388227
View Documents
Thermo Scientific Alfa Aesar
022956.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

12037-42-2
513.64
FTGZPVKRMCXHDZ-UHFFFAOYSA-N
72720435
O=[V]=O.O=[V]=O.O=[V]=O.O=[V]=O.O=[V](=O)O[V](=O)=O
O13V6
MFCD00064848
vanadium oxide v6o13, vanox 13
dioxovanadiooxy(dioxo)vanadium;dioxovanadium

Specifications

∼1500°C
Black-Blue
O13V6
UN3285
Soluble in nitric,hydrofluoric,and concentrated sulfuric acids. Insoluble in water.
O=[V]=O.O=[V]=O.O=[V]=O.O=[V]=O.O=[V](=O)O[V](=O)=O
513.64
513.64
Odorless
(metals basis)
12037-42-2
Crystalline Powder
MFCD00064848
vanadium oxide v6o13, vanox 13
FTGZPVKRMCXHDZ-UHFFFAOYSA-N
dioxovanadiooxy(dioxo)vanadium;dioxovanadium
72720435
99.5%
3.91 g/mL
Vanadium(IV,V) oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c

H302+H332-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.