Learn More
Valeryl chloride, 98%, Thermo Scientific Chemicals
31.30 € - 349.00 €
Chemical Identifiers
CAS | 638-29-9 |
---|---|
Molecular Formula | C5H9ClO |
Molecular Weight (g/mol) | 120.58 |
MDL Number | MFCD00000757 |
InChI Key | XGISHOFUAFNYQF-UHFFFAOYSA-N |
Synonym | valeryl chloride, valeroyl chloride, valerylchloride, n-valeryl chloride, n-pentanoyl chloride, n-valeroyl chloride, pentanoic acid chloride, n-pentoyl chloride, valerylchlorid, valeroylchloride |
PubChem CID | 61186 |
IUPAC Name | pentanoyl chloride |
SMILES | CCCCC(Cl)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11445247
|
Thermo Scientific Alfa Aesar
B22637.18 |
50 g |
31.30 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11455247
|
Thermo Scientific Alfa Aesar
B22637.30 |
250 g |
92.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11435247
|
Thermo Scientific Alfa Aesar
B22637.0B |
1000 g |
349.00 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Valeryl chloride is an intermediate for pharmaceuticals, agrochemicals, surfactants, fragrances, electronic chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsValeryl chloride is an intermediate for pharmaceuticals, agrochemicals, surfactants, fragrances, electronic chemicals.
Solubility
Hydrolyzes in water. Soluble in ether, chloroform.
Notes
Moisture sensitive. Stay away from oxidizing agents, bases, alcohols.
Chemical Identifiers
638-29-9 | |
120.58 | |
XGISHOFUAFNYQF-UHFFFAOYSA-N | |
61186 | |
CCCCC(Cl)=O |
C5H9ClO | |
MFCD00000757 | |
valeryl chloride, valeroyl chloride, valerylchloride, n-valeryl chloride, n-pentanoyl chloride, n-valeroyl chloride, pentanoic acid chloride, n-pentoyl chloride, valerylchlorid, valeroylchloride | |
pentanoyl chloride |
Specifications
638-29-9 | |
125°C to 127°C | |
Acrid | |
MFCD00000757 | |
506293 | |
XGISHOFUAFNYQF-UHFFFAOYSA-N | |
pentanoyl chloride | |
61186 | |
98% |
-110°C | |
32°C (89°F) | |
C5H9ClO | |
UN2502 | |
valeryl chloride, valeroyl chloride, valerylchloride, n-valeryl chloride, n-pentanoyl chloride, n-valeroyl chloride, pentanoic acid chloride, n-pentoyl chloride, valerylchlorid, valeroylchloride | |
CCCCC(Cl)=O | |
120.58 | |
120.58 | |
Valeryl chloride |
Safety and Handling
GHS H Statement
H314-H318-H226-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: VALERYL CHLORIDE
missing translation for 'tsca' : TSCA
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H226-Flammable liquid and vapour.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only