missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valerophenone, 99%, Thermo Scientific Chemicals
43.10 € - 507.00 €
Chemical Identifiers
CAS | 1009-14-9 |
---|---|
Molecular Formula | C11H14O |
Molecular Weight (g/mol) | 162.23 |
MDL Number | MFCD00009480 |
InChI Key | XKGLSKVNOSHTAD-UHFFFAOYSA-N |
Synonym | valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci |
PubChem CID | 66093 |
ChEBI | CHEBI:36812 |
IUPAC Name | 1-phenylpentan-1-one |
SMILES | CCCCC(=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11468793
|
Thermo Scientific Alfa Aesar
A10525.14 |
25 g |
43.10 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478793
|
Thermo Scientific Alfa Aesar
A10525.22 |
100 g |
120.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11488793
|
Thermo Scientific Alfa Aesar
A10525.36 |
500 g |
507.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Valerophenone is used as intermediates of liquid crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsValerophenone is used as intermediates of liquid crystals.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents, acids, bases, plastics. Store at room temperature with proper ventilation.
Chemical Identifiers
1009-14-9 | |
162.23 | |
XKGLSKVNOSHTAD-UHFFFAOYSA-N | |
66093 | |
1-phenylpentan-1-one |
C11H14O | |
MFCD00009480 | |
valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci | |
CHEBI:36812 | |
CCCCC(=O)C1=CC=CC=C1 |
Specifications
1009-14-9 | |
0.978 g/mL | |
102°C (215°F) | |
C11H14O | |
MFCD00009480 | |
valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci | |
XKGLSKVNOSHTAD-UHFFFAOYSA-N | |
1-phenylpentan-1-one | |
66093 | |
162.23 | |
Valerophenone |
°C to 9°C | |
244°C to 245°C | |
99% | |
1.5143 | |
1907717 | |
Insoluble in water. | |
CCCCC(=O)C1=CC=CC=C1 | |
162.23 | |
CHEBI:36812 | |
99% |
Safety and Handling
missing translation for 'einecsNumber' : 213-767-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only