Learn More
Thermo Scientific™ Valco™ Injector Model C6W
Use the Thermo Scientific™ Valco™ Injector Model C6W as an injector or switching valve.
Brand:  Thermo Scientific™ SL5CW
 View more versions of this product
Product Code. 11513990
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Use the Thermo Scientific™ Valco™ Injector Model C6W as an injector or switching valve. Available with manual or a microelectric actuator. Has 1/16 in. fittings and six 0.010 inch ports.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Specifications
Microelectric or Manual
Sample injector loop, stainless steel
6 Ports
5 μL
Injector/Switching Valve
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.