Learn More
Integra Biosciences™ VACUSAFE™ Lid
Specifically designed lid for the VACUSAFE™ aspiration system. Integra Biosciences™ VACUSAFE™ Lid includes integrated tubing connections.
112.00 € - 790.00 €
Specifications
For Use With (Application) 4L VACUSAFE aspiration bottle
Product Type Lid
View More Specs
Products 2
Product Code Brand For Use With (Application) Includes Price Quantity & Availability  
Product Code Brand For Use With (Application) Includes Price Quantity & Availability  
10098601
View Documents Promotion Details
Integra Biosciences™
158372
4L VACUSAFE aspiration bottle Tubing fittings
112.00 €
Each
In Stock
to see stock.
Edge
Add to basket
10193102
View Documents Promotion Details
Integra Biosciences™
158401
4L VACUSAFE aspiration bottle Quick couplings (PVDF) and level detection
790.00 €
Each
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
Description
Description
  • Suitable for the VACUSAFE™ vacuum aspiration system
  • Integrated tubing connections
  • Quick couplings and level detection in some models
Specifications
Specifications
4L VACUSAFE aspiration bottle
Lid