Learn More
Greiner Bio-One VACUETTE™ K3EDTA Blood Collection Tubes
VACUETTE™ EDTA Blood Collection tubes are used for determinations in EDTA whole blood for haematology. Available with K2EDTA, K2EDTA Separator and K3EDTA. VACUETTE™ K3EDTA tubes contain tri-potassium K3EDTA which blocks the coagulation cascade.
Brand: Greiner Bio-One 454021
Description
VACUETTE™ EDTA Blood Collection tubes are used for determinations in EDTA whole blood for haematology. Available with K2EDTA, K2EDTA Separator and K3EDTA. VACUETTE™ K3EDTA tubes contain tri-potassium K3EDTA which blocks the coagulation cascade.
- Tubes are supplied in packs of 50 pieces
- For venous blood collection
- Made out of virtually unbreakable PET plastic
- Improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
- Primarily in hematology
- Inner wall is coated with spray-dried K2EDTA which acts as an anticoagulant, binding calcium ions and interrupting the clotting cascade
- K2EDTA Blood Collection Tubes with Gel Separator and K3EDTA Blood Collection Tubes are available
- Pink caps for immunohematology are also available
- CE marked
Specifications
Blood Collection Tube | |
PET | |
Safety Pull Cap | |
VACUETTE™ Blood Collection Needles | |
75 mm | |
Blood Collection |
K3 EDTA | |
Sterile | |
4 mL | |
Lavender with Black Ring | |
13 x 75 mm | |
13 mm |
Safety and Handling
- vacuette K3EDTA blood collection tube
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Serious eye damage/eye irritation Category 2
- Acute toxicity Category 4
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : EDTA K3
Your input is important to us. Please complete this form to provide feedback related to the content on this product.