missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ V4A Stainless Steel Novartos Uno Pharma Zone Samplers (Triple-Sample)

For Unit-Dose-Sampling. It is now possible to withdraw a triple-sample without inserting the lance repeatedly into the bulk material.

2185.00 € - 2360.00 €

Specifications

Product Type Powder Sampler
Material Stainless Steel
Autoclavable Autoclavable
View More Specs

Products 3
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
17220584
View Documents
Buerkle™
5358-0630
600 mm
2185.00 €
Each
In Stock
to see stock.
Add to basket
 
17240584
View Documents
Buerkle™
5358-1030
1000 mm
2285.00 €
Each
In Stock
to see stock.
Add to basket
 
17250584
View Documents
Buerkle™
5358-1430
1400 mm
2360.00 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • The sample inserts fit precisely and can easily be replaced
  • Inserts are available in volumes from 0 to 4.0mL
  • The insertion depths can easily be read from the laser-engraved depth scale
  • Stainless steel AISI 316L (1.4404) / AISI 316Ti (1.4571)
  • Groove-free and hand-polished
  • 25 mm Ø
Specifications

Specifications

Powder Sampler
Stainless Steel
Autoclavable
Videos
SDS
Documents

Documents

Product Certifications

Please order inserts separately!

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.