Learn More
Uvex™ C300 Foam Cut Protection Gloves
Click to view available options
Size:
7
8
9
10
Description
- Gloves provide level 3/C blade cut resistance, abrasion resistance, and high tear resistance
- High-performance elastomer (HPE) coating on the palm and fingers for enhanced grip capabilities
- Soft-grip foam coating provides mechanical abrasion resistance
- Highly flexible with very good tactile sensation
- Uvex Climazone technology reduces excessive moisture and warmth
- 3D Ergo technology for a secure fit
- Silicone-free according to the imprint test
Specifications
Specifications
Coating Material | HPE |
Color | Gray |
Material | Bamboo Viscose, HPPE, Polyamide, Glass |
Size | 8 |
EN388 Standard | 3X42C |
Glove Cuff Style | Knit |
Series | C300 Foam |