Learn More
Vilber Lourmat™ UV Lamp Stand
Rich and robust table stand for ultraviolet lamps 4/8 W.
Brand:  Vilber Lourmat™ 9130-2001-1
 View more versions of this product
Product Code. 11850240
  • 90.50 € / Each
Explore more special offers
Description
Description
  • Stand for UV lamps 4/8W
  • Double wavelength lamp
  • 254/365 nm Intensity 
  • Lamp 2 x 6 W
  • M, 2 table stand
Specifications
Specifications
4/8w Handheld UV Lamp
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Vilber Lourmat™ UV Lamp Stand > For Use With: 4/8w Handheld UV Lamp