Learn More
UNC13A/Munc13-1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55053-1-AP-150UL
 View more versions of this product
Product Code. 16826495
  • 399.00 € / 150µL
Explore more special offers
Description
Description
The antibody is specific to UNC13A.
UNC13A, also named as KIAA1032, belongs to the unc-13 family. It plays a role in vesicle maturation during exocytosis as a target of the diacylglycerol second messenger pathway. UNC13A is involved in neurotransmitter release by acting in synaptic vesicle priming prior to vesicle fusion and participates in the activity-dependent refilling of readily releasable vesicle pool (RRP). It is essential for synaptic vesicle maturation in most excitatory/glutamatergic but not inhibitory/GABA-mediated synapses. UNC13A is also involved in secretory granule priming in insulin secretion.
Specifications
Specifications
UNC13A/Munc13-1
Polyclonal
Unconjugated
UNC13A
KIAA1032, Munc13 1, Protein unc 13 homolog A, unc 13 homolog A (C. elegans), UNC13A
Rabbit
Antigen Affinity Chromatography
RUO
23025, 382018
-20°C
Liquid
Immunoprecipitation, Western Blot
0.33 mg/mL
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4KUS2, Q9UPW8
Unc13a
Peptide
150 μL
Primary
Human, Mouse