missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ U-D Doser

Ideal for accurately sampling specific volumes of liquids. Sampling Systems™ U-D Doser is designed specifically for unit dose sampling of liquids in the pharmaceutical industry.

Brand:  Sampling Systems™ 5010A-600

Product Code. 15289747

  • 4240.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sampler body 316L Stainless Steel
  • Sample chamber is designed so that the outside of the syringe is not contaminated during sampling
  • The syringe can therefore be used as a carrying container for taking samples back to the laboratory
Specifications

Specifications

Stainless Steel (316L)
5 mL
600 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.