missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 800 J

Product Code. 15864993 Shop All DuPont Products
Medium
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Tyvek™ 800 J coveralls benefit from the use of a specific 'impervious technology' to achieve an effective barrier against many low-concentrated water-based inorganic chemicals (even under pressure) and small-sized hazardous particulates, as well as oil repellency.

  • Tyvek™ 800J garments combine resistance to low-concentration, water-based, inorganic chemicals (even under pressure) with the strength and durability of Tyvek™ thanks to an innovative fabric technology and enhanced garment design. Developed to our highest standards, Tyvek™ 800J coveralls are soft, comfortable and lightweight without compromising their level of durability. They are now available with serged and over-taped seams that provide the performance of Type 3/4/5/6 garments, which have been tested to standards against pressurized liquids, heavy liquid aerosols and airborne solid particles. The coveralls provide an effective barrier against many water-based inorganic chemicals in low concentration and particles (down to 1.0 micron in size), as well as oil repellency. Tyvek™ 800J coveralls are suitable for applications such as industrial cleaning, chemical packaging and redistribution, waste treatment and disposal, and environmental remediation. Tyvek™ 800J Type 3/4/5/6 coveralls offer the following safety and comfort benefits:
    • Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
    • Protection against infective agents (EN 14126) including resistance to penetration by blood and body fluids using synthetic blood (ISO 16603) and resistance to penetration by blood-borne pathogens using Phi-X174 bacteriophage (ISO 16604)
    • EN 1073-2 (protection against radioactive contamination)
    • Fabric and seams offer chemical permeation barrier to low concentration water-based inorganic chemicals
    • Serged and over-taped seams for protection and strength
    • Self-adhesive chin flap for tight seal of suit to the mask
    • Elastic face, wrists and ankles as well as glued-in waist elastic
    • Elastic thumb loops keep sleeves in place
TRUSTED_SUSTAINABILITY

Specifications

Size Medium
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Waist Style Elastic (Glued-in)
Ankle Style Elastic
Cuff Style Thumb Loop, Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Front Zipper with Flap
Seam Type Stitched, Taped
Protection Class Category III, Type 3-B, 4-B, 5-B and 6-B EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
Resistance Chemicals, Liquid, Particle, Anti-static, Water
Collar Type Hooded
Certifications/Compliance Certified according to Regulation (EU) 2016/425 Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination) Antistatic treatment (EN 1149-5) - on inside
Description Tyvek™ 800 J
No. of Pockets None
Size (English) Chest 36-39 in.
Size (Metric) Chest 92 to 100 cm
Thickness 160 μm
For Use With (Application) Industrial cleaning, work at petrochemical installations, in sewers and maintenance operations.
Height (Metric) 168 to 176 cm
Weight (English) 59 g/m2
Quantity 25/Cs.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuPont™ Tyvek™ 800 J >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.