missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ 500 Lab Coats
Tyvek™ Lab coats offer enhanced protection for body parts that are more exposed to hazardous substances.
373.00 €
Specifications
Product Type | Chemical Resistant Lab Coat |
---|---|
Gender | Unisex |
Color | White |
Closure Type | Zipper |
Cuff Style | Elastic |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
16367381
|
DuPont™
D15534010 |
Small |
373.00 €
Pack of 50 |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
16377381
|
DuPont™
D15534011 |
Medium |
373.00 €
Pack of 50 |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
16387381
|
DuPont™
D15534012 |
Large |
373.00 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
16397381
|
DuPont™
D15534014 |
2X-Large |
373.00 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
Description
- Composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort
- Permeable to both air and water vapour
- Offers an excellent barrier against fine particles and fibres (down to 1 micron in size)
- Certified according to Regulation (EU) 2016/425
- Partial body chemical protective clothing, Category III, Type PB (6-B)
- EN 14126 (barrier to infective agents)
- Antistatic treatment (EN 1149-5) on both sides
- Applications include: pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting, and general clean-up
Specifications
Chemical Resistant Lab Coat | |
White | |
Elastic | |
Non-sterile | |
Stitched |
Unisex | |
Zipper | |
Limited-use | |
Chemicals |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title