Learn More
DuPont™ Tyvek™ 500 HV Coveralls
Product Code. 15682884
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Offer an excellent barrier against fine particles and fibres. DuPont™ Tyvek™ 500 HV Coveralls are composed of flash spun polyethylene, providing an ideal balance of protection, durability and comfort.
  • Robust yet lightweight
  • Mandarin collar, elasticated wrists and ankles as well as glued-in elasticated waist.
  • Ample crotch area for freedom of movement
  • Large, easy-to grasp zipper puller
  • Composed of flash spun polyethylene, providing an ideal balance of protection, durability and comfort
  • Permeable to both air and water vapour, yet repel water-based liquids and aerosols
  • Ultra-low linting and antistatically treated on the inside
  • Chemical protective clothing, Category III, Type 5-B and 6-B EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), EN ISO 20471 (High Visibility Clothing, Class 3) Antistatic treatment (EN 1149-5) on the inside
  • Stitched external seams
  • Optimised design provides very low inward leakage thanks toTyvek™ auto-lock zipper and zipper flap for increased protection
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large
Material Polyethylene
Color Fluorescent Orange, Reflective Silver
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Seam Type Stitched
Collar Type Mandarin
Certifications/Compliance Chemical protective clothing, Category III, Type 5-B and 6-B, EN 14126, EN 1073-2, EN ISO 20471, EN 1149-5 (inside),
Description Nonwoven, flash spun construction; Fluorescent orange with silver reflective bands