missing translation for 'onlineSavingsMsg'
Learn More

Tyramine hydrochloride, 98%, Thermo Scientific Chemicals

30.50 € - 116.00 €

Chemical Identifiers

CAS 60-19-5
Molecular Formula C8H12ClNO
Molecular Weight (g/mol) 173.64
MDL Number MFCD00012901
InChI Key RNISDHSYKZAWOK-UHFFFAOYSA-N
Synonym tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
PubChem CID 66449
IUPAC Name 4-(2-aminoethyl)phenol;hydrochloride
SMILES [H+].[Cl-].NCCC1=CC=C(O)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11484893
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12220.06
5 g
30.50 €
5g
Please to purchase this item. Need a web account? Register with us today!
11494893
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12220.14
25 g
116.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Reagent for fluorescence enzyme immunoassay

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

60-19-5
173.64
RNISDHSYKZAWOK-UHFFFAOYSA-N
66449
[H+].[Cl-].NCCC1=CC=C(O)C=C1
C8H12ClNO
MFCD00012901
tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
4-(2-aminoethyl)phenol;hydrochloride

Specifications

60-19-5
C8H12ClNO
3627058
tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
[H+].[Cl-].NCCC1=CC=C(O)C=C1
173.64
173.65
Tyramine hydrochloride
271°C to 274°C
MFCD00012901
14,9835
RNISDHSYKZAWOK-UHFFFAOYSA-N
4-(2-aminoethyl)phenol;hydrochloride
66449
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 200-462-5

missing translation for 'rtecsNumber' : SJ6050000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only