missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-PHOENIX Touchless-90 Light Gray
Safe and approved storage of flammable hazardous substances in working areas
3213.92 € - 5092.77 €
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant |
---|---|
Color | Light Gray |
Material | Steel |
Product Type | Safety storage cabinet |
For Use With (Application) | Safe and approved storage of flammable hazardous substances in working areas |
Alert:
For indoor use only
Product Code | Brand | Width (Metric) Exterior | Depth (Metric) Exterior | Configuration | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) Exterior | Depth (Metric) Exterior | Configuration | Price | Quantity & Availability | |||
15913736
|
asecos™
3023700133659 |
596 mm | 616 mm | 3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) |
3213.92 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15953736
|
asecos™
3023700133663 |
596 mm | 616 mm | 3 x tray shelf, 1 x perforated insert (320 mm), 1 x bottom collecting sump (sheet steel powder-coated) |
3415.32 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15963736
|
asecos™
3023700133681 |
596 mm | 616 mm | 4 x drawer (sheet steel powder-coated) |
3564.78 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15983736
|
asecos™
3023700133683 |
596 mm | 616 mm | 6 x drawer (sheet steel powder-coated) |
3844.62 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15914266
|
asecos™
3031700133695 |
596 mm | 749 mm | 4 x drawer (sheet steel powder-coated) |
3767.24 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15934266
|
asecos™
3031700133697 |
596 mm | 749 mm | 6 x drawer (sheet steel powder-coated) |
4087.36 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15902726
|
asecos™
3012000133715 |
1196 mm | 616 mm | 3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) |
4007.86 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15942726
|
asecos™
3012000133719 |
1196 mm | 616 mm | 3 x tray shelf, 1 x perforated insert (365 mm), 1 x bottom collecting sump (sheet steel powder-coated) |
4210.32 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15992726
|
asecos™
3012000133724 |
1196 mm | 616 mm | 4 x drawer (sheet steel powder-coated) |
4649.69 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15912736
|
asecos™
3012000133726 |
1196 mm | 616 mm | 6 x drawer (sheet steel powder-coated) |
5092.77 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Space saving folding doors: less required space in front of the cabinet while doors are open
- Effortless use: the sensor detects a person within approx. 2 seconds
- User-friendly and safe: after detection, the folding doors open automatically within approx. 5 seconds. The sensor recognises the person while operating and hold the folding doors open
- Always closed: if the person leaves the sensor-controlled detection area, the folding doors close automatically after approx. 30 seconds
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant | |
Steel | |
Safe and approved storage of flammable hazardous substances in working areas | |
1742 mm |
Light Gray | |
Safety storage cabinet | |
1968 mm | |
Powder-coated plain |