missing translation for 'onlineSavingsMsg'
Learn More
DuPont™ Tychem™ NT450 Product Code.: 16370466

DuPont™ Tychem™ NT450

Product Code. 16370466 Shop All DuPont Products
Small, 8
Click to view available options
Size:
Large, 10
Medium, 9
Small, 8
X-Large, 11
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 16370466

Brand: DuPont™ D15536299

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Flexible impermeable knit-lined dual nitrile glove. Ideal for heavy duty work or longer exposure to hazardous chemicals and solvents.

  • Double nitrile coated gloves for jobs requiring chemical performance and comfort, plus excellent adherence
  • Lightweight,'Second-Skin' feel and low-modulus formulation reduces hand fatigue
  • CE CAT III - EN388: 4112X, EN ISO 374-1:2016/TYPE A, JKLOPT
  • EN ISO 374-5:2016
  • Featuring a cotton/polyester liner that absorbs perspiration for added comfort and a rough textured finish that ensures excellent grip
  • Food contact compatible
  • Resists Hydrochloric Acid (37%) - Level 6 (breakthrough time EN374)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) CE Cat III
Color Blue
Exterior Finish Rough
Latex Free No
Material Nitrile
Size Small, 8
Style Scalloped edge / Anatomical shape
Thickness (English) 56 mil
Thickness (Metric) 1.42 mm
EN388 Standard EN388: 4112X
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLOPT
EN ISO 374 - 5 Yes
Glove Cuff Style Plain
Liner Material Cotton/polyester liner
Lining Lined
Model NT450
Quantity 12 pairs per bag/6 bags per box: 72 total pairs
Resistant To Resists Hydrochloric Acid (37%) - Level 6 (breakthrough time EN374)
Length (English) 14 in.
Length (Metric) 356 mm
Show More Show Less

  1. Store away from light and humidity
  2. Rinse gloves in running water before removing, using a neutral detergent if necessary
  3. Gloves should not be used around heat, open flame, sparks or in potentially flammable environments
  4. For Professional Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuPont™ Tychem™ NT450 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.