Learn More
DuPont™ Tychem™ 6000 F FaceSeal
Product Code. 16358555
Click to view available options
Size:
Small
Medium
Large
2X-Large
3X-Large
4X-Large
5X-Large
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Tychem™ 6000 F FaceSeal is a hooded coverall that includes a rubber seal for a tight fit.
  • Made from a lightweight and durable fabric (< 500 g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek™ substrate
  • Offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards, and certain chemical warfare agents
  • Certified according to Regulation (EU) 2016/425
  • Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
  • EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
  • Antistatic treatment (EN 1149-5) on inside
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Back entry with zipper, double zipper flap with snaps and hook and loop closure
  • Attached undergloves
  • Used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Size 2X-Large
Ankle Style Attached socks
Closure Type Rear-entry zipper with double flap, snaps and hook-and-loop seal
Cuff Style Attached gloves
Disposable Single-use
Gender Unisex
Neck Style Hooded
Quantity 1/Ea.
Seam Type Stitched, Taped
Sterility Non-sterile
Waist Style Elastic