missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 2000 C with Socks
Shop All DuPont Products
Click to view available options
Size: Small
Description
- Made from a lightweight and durable fabric (< 450 g per garment)
- Offers good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure)
- Used for splash or pressurised splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing
- Certified according to Regulation (EU) 2016/425
- Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
- EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
- Antistatic treatment (EN 1149-5) on inside
- Stitched and over-taped seams with barrier tape for protection and strength

Specifications
Specifications
Size | Small |
Gender | Unisex |
Color | Yellow |
Disposable | Single-use |
Waist Style | Elastic |
Ankle Style | Attached Socks, Boot Overflaps, Elastic |
Cuff Style | Thumb Loop |
Neck Style | Hooded |
Closure Type | Double Zipper with Self-adhesive Flap and Chin Flap |
Seam Type | Stitched, Taped |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DuPont™ Tychem™ 2000 C with Socks >
Spot an opportunity for improvement?Share a Content Correction