Learn More
TUSC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11538-1-AP-150UL
 View more versions of this product
Product Code. 16848683
  • 359.47 € / 150µL
Explore more special offers
Description
Description
TUSC2 (tumor suppressor candidate 2), also known as FUS1, LGCC or PDAP2, is a 110 amino acid protein that is expressed at high levels in kidney, pancreas, lung, heart and skeletal muscle where it is thought to function as a tumor suppressor. More specifically, TUSC2 inhibits invasive colony formation by inducing G1 cell cycle arrest and ultimately causing apoptosis, thereby preventing tumor growth. The gene encoding TUSC2 maps to human chromosome 3, which houses over 1,100 genes, including a chemokine receptor (CKR) gene cluster and a variety of human cancer-related gene loci. Key tumor suppressing genes on chromosome 3 include those that encode the apoptosis mediator RASSF1, the cell migration regulator HYAL1 and the angiogenesis suppressor SEMA3B.
Specifications
Specifications
TUSC2
Polyclonal
Unconjugated
TUSC2
C3orf11, Fus 1 protein, FUS1, Fusion 1 protein, LGCC, PAP, PDAP2, PDGFA associated protein 2, tumor suppressor candidate 2, TUSC2
Rabbit
Antigen Affinity Chromatography
RUO
11334, 501052, 80385
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75896, Q9WVF8
Tusc2
TUSC2 Fusion Protein Ag2118
150 μL
Primary
Human, Mouse, Rat