Learn More
Fisherbrand™ Turbo PTFE Stir Bar Retriever with Rare Earth Core
Fisherbrand™ Turbo PTFE Stir Bar Retriever with Rare Earth Core are used to retrieve stir bars from mixing containers. The PTFE stir bar retriever is chemically resistant, inert, and leak-free. Uses Samarium-Cobalt magnets at its core.
Brand:  Fisherbrand™ 004.2350.RE
 View more versions of this product
Product Code. 16235871
  • 56.80 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Inert and leak-free
  • Molded in pure, FDA-approved PTFE
  • Alnico V magnets
  • Autoclavable
  • For girant stirrer bars
Specifications
Specifications
White
For the removal of Giant Stir Bars from vessels of all kinds
350 mm
1
Turbo PTFE Stir Bar Retriever with Rare Earth Core for Giant Stir Bars
18 mm
PTFE with Rare Earth Magnet Core