missing translation for 'onlineSavingsMsg'
Learn More

Tungsten wire basket; # turns, 8; ID(mm), 9; Ht(mm), 14; Wire leads(cm), 4.0; Wire dia(mm), 0.76, Thermo Scientific™

Product Code. 15484845
Click to view available options
Quantity:
10 Pc.
50 Pc.
This item is not returnable. View return policy
This item is not returnable. View return policy

For vacuum deposition

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Quantity 50 Pc.
Form Wire
Chemical Name or Material Tungsten wire basket, 8 turns, ID(mm), 9; Ht(mm), 14; Wire leads(cm), 4.0; Wire dia(mm), 0.76
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'