missing translation for 'onlineSavingsMsg'
Learn More

Tungsten(VI) oxide, Puratronic™, 99.998% (metals basis excluding Mo), Mo 100ppm, Thermo Scientific Chemicals

176.88 € - 580.89 €

Chemical Identifiers

CAS 1314-35-8
Molecular Formula O3W
Molecular Weight (g/mol) 231.837
MDL Number MFCD00011466
InChI Key ZNOKGRXACCSDPY-UHFFFAOYSA-N
Synonym tungsten trioxide, tungsten vi oxide, tungstic oxide, tungstic anhydride, tungsten oxide wo3, tungsten blue, tungstic acid anhydride, wolframic acid, anhydride, tungstentrioxide, acmc-20ajaa
PubChem CID 14811
IUPAC Name trioxotungsten
SMILES O=[W](=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11336718
SDS View Documents
Thermo Scientific Alfa Aesar
013398.14
25 g
176.88 €
25g
Please to purchase this item. Need a web account? Register with us today!
11326718
SDS View Documents
Thermo Scientific Alfa Aesar
013398.22
100 g
580.89 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tungsten(VI) oxide is used in tungsten and tungstate manufacturing which are used as X-ray screens and for fire proofing fabrics. It is used as a ceramic pigment. Nanowires of tungsten(VI) oxide are capable of absorbing a higher percentage of the sun radiation since it absorbs blue light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-231-4

missing translation for 'rtecsNumber' : YO7760000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tungsten(VI) oxide, Puratronic™, 99.998% (metals basis excluding Mo), Mo 100ppm, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.