missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten(V) ethoxide, Thermo Scientific Chemicals
Intermediate for sol-gel coatings
101.00 € - 393.00 €
Chemical Identifiers
CAS | 26143-11-3 |
---|---|
Molecular Formula | C10H25O5W |
Molecular Weight (g/mol) | 409.15 |
MDL Number | MFCD00078041 |
InChI Key | SRGJJDPFERQMIL-UHFFFAOYSA-N |
Synonym | tungsten v ethoxide, pentaethoxywolfram v, tungsten pentaethoxide, acmc-1cbw9, tungsten 5+ pentakis ethoxide |
PubChem CID | 57357833 |
IUPAC Name | ethanolate;tungsten |
SMILES | CCO[W](OCC)(OCC)(OCC)OCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11318958
|
Thermo Scientific Alfa Aesar
041794.04 |
2 g |
101.00 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11328958
|
Thermo Scientific Alfa Aesar
041794.09 |
10 g |
393.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tungsten(V) ethoxide is used as an intermediate for sol-gel coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
26143-11-3 | |
409.15 | |
SRGJJDPFERQMIL-UHFFFAOYSA-N | |
57357833 | |
CCO[W](OCC)(OCC)(OCC)OCC |
C10H25O5W | |
MFCD00078041 | |
tungsten v ethoxide, pentaethoxywolfram v, tungsten pentaethoxide, acmc-1cbw9, tungsten 5+ pentakis ethoxide | |
ethanolate;tungsten |
Specifications
26143-11-3 | |
>110°C (230°F) | |
W(OC2H5)5 | |
Moisture sensitive | |
Soluble in hydrocarbons. | |
CCO[W](OCC)(OCC)(OCC)OCC | |
409.15 | |
409.15 | |
Tungsten(V) ethoxide |
110°C to 115°C (1 mmHg) | |
C10H25O5W | |
MFCD00078041 | |
tungsten v ethoxide, pentaethoxywolfram v, tungsten pentaethoxide, acmc-1cbw9, tungsten 5+ pentakis ethoxide | |
SRGJJDPFERQMIL-UHFFFAOYSA-N | |
ethanolate;tungsten | |
57357833 | |
Liquid |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only