missing translation for 'onlineSavingsMsg'
Learn More
Tungsten carbide Cobalt, 99% (metals basis), Thermo Scientific Chemicals Product Code.: 11358227

Tungsten carbide Cobalt, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11358227
250 g
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11358227

Brand: Thermo Scientific Alfa Aesar 022955.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tungsten carbide Cobalt is used in cutting tools, having hard ceramic crystals of a few microns in size within a ductile metal matrix, use as electrodes for Electrode Discharge Machining. It is also used in production of tools for milling, drilling and pressing/punching processes, surgical instruments, product is the rotating balls used in ball point pens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12774-15-1
Molecular Formula CHCoW
Molecular Weight (g/mol) 255.79
MDL Number MFCD00064847
InChI Key IKEHVRMRUVYPGG-UHFFFAOYSA-N
Synonym tungsten carbide cobalt, methylidynetungsten cobalt
PubChem CID 56846058
SMILES [Co].C#[W]

Specifications

Color Gray
Avg. Mol. Wt. or Mol. Wt. Range WC:Co; 94:6 wt%
UN Number UN3178
Quantity 250 g
Solubility Information Insoluble in water.
IUPAC Name cobalt;methylidynetungsten
Percent Purity 99%
Mesh Size −325 Mesh
Odor Odorless
Form Powder
Assay (metals basis)
Chemical Name or Material Tungsten carbide Cobalt
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tungsten carbide Cobalt, 99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.