missing translation for 'onlineSavingsMsg'
Learn More

Tungsten carbide, 99.5% (metals basis), Thermo Scientific Chemicals

191.00 € - 706.00 €

Chemical Identifiers

CAS 12070-13-2
Molecular Formula CW2
Molecular Weight (g/mol) 379.69
MDL Number MFCD00801036
InChI Key FHLFYDOXSZQEPO-UHFFFAOYSA-N
Synonym tungsten carbide w2c, methyliumylidyneditungstenene
PubChem CID 102601578
IUPAC Name methyliumylidyneditungstenene
SMILES [W-]=[W]#[C+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11388808
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041665.18
50 g
191.00 €
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11378808
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041665.30
250 g
706.00 €
250g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tungsten carbide is used in industrial machinery, cutting tools, abrasives, armor-piercing rounds and jewelry. It is preferred even over stainless steel because of their incredible hardness and resistance to wear and tear. It is utilized for the manufacture of mill products including grinding and milling. It also includes tipping trekking, ski poles as well as cleats. It is mainly used in the form of cemented tungsten carbide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12070-13-2
379.69
FHLFYDOXSZQEPO-UHFFFAOYSA-N
102601578
[W-]=[W]#[C+]
CW2
MFCD00801036
tungsten carbide w2c, methyliumylidyneditungstenene
methyliumylidyneditungstenene

Specifications

∼2800°C
Black
CW2
MFCD00801036
tungsten carbide w2c, methyliumylidyneditungstenene
FHLFYDOXSZQEPO-UHFFFAOYSA-N
methyliumylidyneditungstenene
102601578
99.5%
(metals basis)
12070-13-2
Powder
W2C
UN3178
Soluble in nitric acid and hydrofluoric acid. Insoluble in water.
[W-]=[W]#[C+]
379.69
379.71
17.15 g/mL
Tungsten carbide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P280-P370+P378q

H228

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 235-124-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only