missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Tube Block

Product Code. 15782549
Click to view available options
Holds:
Conical Tubes
Cryotubes
Eppendorf Tubes
Tubes
No. of Places:
4
8
9
24
30
Tube Size:
0.5 mL
1.5 mL
15 mL
2 mL
5 mL
5 to 7 mL
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

OHAUS™ Tube Block is constructed of Valox 357 Plastic and accommodates a variety of tubes.

  • Designed to securely hold tubes in place
  • Applications include: cell cultures, bacterial and yeast cultures, immunoassays, protein studies, blotting techniques, staining and destaining gels. solubility studies, extraction procedures, DNA, RNA, hybridizations, enzyme reactions, ELISA assays
TRUSTED_SUSTAINABILITY

Specifications

Description Block For 4 X 50 mL Conical Tubes
For Use With (Equipment) Shaker
Holds Conical Tubes
Material Valox 357 Plastic
No. of Places 4
Type Block
Tube Size 50 mL
Dimensions (L x W) 10.20 cm x 14.50 cm
Dimensions (L x W x H) 10.2 x 14.5 x 12.2 cm
Length (English) 4.00 in.
Length (Metric) 10.20 cm
Width (English) 5.70 in.
Width (Metric) 14.50 cm
Height (English) 4.80 in.
Height (Metric) 12.200 cm
Depth (English) 4.00 in.
Depth (Metric) 10.20 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'