missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBB3 Mouse anti-Human, Mouse, Rat, Clone: 1F8G10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66375-1-IG-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to TUBB3 but not cross-react with other tubulin isoforms.
The betaIII-tubulin isoform is present domitly in cells of neuronal origin and it is one of the earliest markers of neuronal differentiation. It is regarded as a specific probe for the cells of neuronal origin as well as for the tumours originating from these cells. The betaIII-tubulin is most abundant in cells of neuronal origin, but was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues.Specifications
TUBB3 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13509, Q4QRB4, Q9ERD7 | |
TUBB3 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
1F8G10 | |
Unconjugated | |
TUBB3 | |
beta 4, beta tubulin, MC1R, TUBB, TUBB3, TUBB3-specific, Tubulin beta 3 chain, Tubulin beta III, tubulin, beta 3, Tuj1 | |
Mouse | |
Protein G | |
RUO | |
10381, 22152, 246118 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction