Learn More
TSG101 Mouse anti-Human, Unconjugated, Clone: 2B7G8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67381-1-IG-20UL
This item is not returnable.
View return policy
Description
The TSG101 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TSG101. This antibody recognizes human, mouse, rat antigen. The TSG101 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
TSG101 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
2B7G8 | |
Unconjugated | |
20 μL | |
7251 | |
IgG1 |