missing translation for 'onlineSavingsMsg'
Learn More

Tryptamine, 98%, Thermo Scientific Chemicals

29.75 € - 322.61 €

Chemical Identifiers

CAS 61-54-1
Molecular Formula C10H12N2
Molecular Weight (g/mol) 160.22
MDL Number MFCD00005661
InChI Key APJYDQYYACXCRM-UHFFFAOYSA-N
Synonym tryptamine, 2-1h-indol-3-yl ethanamine, 3-2-aminoethyl indole, 1h-indole-3-ethanamine, 2-3-indolyl ethylamine, indol-3-ethylamine, indole, 3-2-aminoethyl, 2-indol-3-yl ethylamine, 2-1h-indol-3-yl ethan-1-amine, tryptamin
PubChem CID 1150
ChEBI CHEBI:16765
IUPAC Name 2-(1H-indol-3-yl)ethanamine
SMILES NCCC1=CNC2=CC=CC=C12
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10720891
View Documents
Thermo Scientific Acros
186120100
10 g Glass bottle
29.75 €
10g
Please to purchase this item. Need a web account? Register with us today!
10427390
View Documents
Thermo Scientific Acros
186120500
50 g Glass bottle
96.08 €
50g
Please to purchase this item. Need a web account? Register with us today!
11996611
View Documents
Thermo Scientific Acros
186122500
250 g Glass bottle
322.61 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

61-54-1
160.22
APJYDQYYACXCRM-UHFFFAOYSA-N
1150
2-(1H-indol-3-yl)ethanamine
C10H12N2
MFCD00005661
tryptamine, 2-1h-indol-3-yl ethanamine, 3-2-aminoethyl indole, 1h-indole-3-ethanamine, 2-3-indolyl ethylamine, indol-3-ethylamine, indole, 3-2-aminoethyl, 2-indol-3-yl ethylamine, 2-1h-indol-3-yl ethan-1-amine, tryptamin
CHEBI:16765
NCCC1=CNC2=CC=CC=C12

Specifications

61-54-1
White
185°C
97.5% min. (GC)
MFCD00005661
15,9976
Solubility in water: negligible. Other solubilities: soluble in ethanol and acetone, practically insoluble in ether, benzene and, chloroform
NCCC1=CNC2=CC=CC=C12
160.22
CHEBI:16765
98%
Tryptamine
113.0°C to 117.0°C
137.0°C (0.1 mmHg)
Authentic
C10H12N2
22,I,636
tryptamine, 2-1h-indol-3-yl ethanamine, 3-2-aminoethyl indole, 1h-indole-3-ethanamine, 2-3-indolyl ethylamine, indol-3-ethylamine, indole, 3-2-aminoethyl, 2-indol-3-yl ethylamine, 2-1h-indol-3-yl ethan-1-amine, tryptamin
APJYDQYYACXCRM-UHFFFAOYSA-N
2-(1H-indol-3-yl)ethanamine
1150
160.22
Crystalline Powder or Flakes
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 200-510-5

Product Identifier
  • Tryptamine

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.