Learn More
Tropolone, 98%, Thermo Scientific Chemicals
Sensitive reagent for reducing sugars. A non-benzenoid aromatic compound, Reagent for the preparation of fused heterocycles and complexes of Ga(III) and In(III).
42.00 € - 418.00 €
Chemical Identifiers
CAS | 533-75-5 |
---|---|
Molecular Formula | C7H6O2 |
Molecular Weight (g/mol) | 122.123 |
MDL Number | MFCD00004158 |
InChI Key | MDYOLVRUBBJPFM-UHFFFAOYSA-N |
Synonym | tropolone, purpurocatechol, 2-hydroxycyclohepta-2,4,6-trienone, 2,4,6-cycloheptatrien-1-one, 2-hydroxy, 2-hydroxytropone, 2-hydroxy-2,4,6-cycloheptatrien-1-one, 2-hydroxy-2,4,6-cycloheptatrienone, unii-7l6dl16p1t, ccris 6609, tropomyosins |
PubChem CID | 10789 |
ChEBI | CHEBI:79966 |
IUPAC Name | 2-hydroxycyclohepta-2,4,6-trien-1-one |
SMILES | C1=CC=C(C(=O)C=C1)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11413684
|
Thermo Scientific Alfa Aesar
A11299.03 |
1 g |
42.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423684
|
Thermo Scientific Alfa Aesar
A11299.06 |
5 g |
122.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11433684
|
Thermo Scientific Alfa Aesar
A11299.14 |
25 g |
418.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tropolone is a sensitive reagent for reducing sugars. A non-benzenoid aromatic compound, Reagent for the preparation of fused heterocycles and complexes of Ga(III) and In(III).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
533-75-5 | |
122.123 | |
MDYOLVRUBBJPFM-UHFFFAOYSA-N | |
10789 | |
2-hydroxycyclohepta-2,4,6-trien-1-one |
C7H6O2 | |
MFCD00004158 | |
tropolone, purpurocatechol, 2-hydroxycyclohepta-2,4,6-trienone, 2,4,6-cycloheptatrien-1-one, 2-hydroxy, 2-hydroxytropone, 2-hydroxy-2,4,6-cycloheptatrien-1-one, 2-hydroxy-2,4,6-cycloheptatrienone, unii-7l6dl16p1t, ccris 6609, tropomyosins | |
CHEBI:79966 | |
C1=CC=C(C(=O)C=C1)O |
Specifications
533-75-5 | |
115°C (12 mmHg) | |
Glass bottle | |
MFCD00004158 | |
Hygroscopic | |
Soluble in water. | |
C1=CC=C(C(=O)C=C1)O | |
122.123 | |
CHEBI:79966 | |
98% | |
Tropolone |
50°C to 55°C | |
>110°C (230°F) | |
C7H6O2 | |
1904978 | |
tropolone, purpurocatechol, 2-hydroxycyclohepta-2,4,6-trienone, 2,4,6-cycloheptatrien-1-one, 2-hydroxy, 2-hydroxytropone, 2-hydroxy-2,4,6-cycloheptatrien-1-one, 2-hydroxy-2,4,6-cycloheptatrienone, unii-7l6dl16p1t, ccris 6609, tropomyosins | |
MDYOLVRUBBJPFM-UHFFFAOYSA-N | |
2-hydroxycyclohepta-2,4,6-trien-1-one | |
10789 | |
122.12 | |
Crystalline |
Safety and Handling
missing translation for 'einecsNumber' : 208-577-2
missing translation for 'rtecsNumber' : GU4075000
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only