Learn More
Thermo Scientific Chemicals Triton™ X-114
65.73 € - 194.97 €
Chemical Identifiers
CAS | 9002-93-1 |
---|---|
Molecular Formula | C16H26O2 |
Molecular Weight (g/mol) | 250.38 |
MDL Number | MFCD00132505 |
InChI Key | JYCQQPHGFMYQCF-UHFFFAOYSA-N |
Synonym | Polyoxyethylene(8) octylphenyl ether |
PubChem CID | 5590 |
IUPAC Name | 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol |
SMILES | CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10537372
|
Thermo Scientific Chemicals
422365000 |
500 mL | Glass bottle |
65.73 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10637754
|
Thermo Scientific Chemicals
422360025 |
2.5 L | Glass bottle |
194.97 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9002-93-1 | |
250.38 | |
JYCQQPHGFMYQCF-UHFFFAOYSA-N | |
5590 | |
CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1 |
C16H26O2 | |
MFCD00132505 | |
Polyoxyethylene(8) octylphenyl ether | |
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol |
Specifications
Undesignated | |
Liquid | |
22° to 28°C (1% in aq. soln.) | |
6 (5% aq. soln.) | |
Authentic | |
1.4880 to 1.4950 | |
C16H26O2 | |
MFCD00132505 | |
Polyoxyethylene(8) octylphenyl ether | |
JYCQQPHGFMYQCF-UHFFFAOYSA-N | |
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol | |
5590 | |
Triton™ X-114 |
9002-93-1 | |
>200.0°C | |
1.0550g/mL | |
254°C | |
pour point: -9°C | |
1.055 | |
260 mPa.s (25°C) | |
15, 6850 | |
Solubility in water: soluble. Other solubilities: miscible in most polar organic solvents and, aromatic hydrocarbons,, insoluble in aliphatic hydrocarbons. | |
CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1 | |
250.38 | |
537 |
Safety and Handling
GHS P Statement
Causes serious eye damage.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'rtecsNumber' : MD0907600
missing translation for 'tsca' : TSCA
- Triton]TS X-114
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.