missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triton™ X-100, Thermo Scientific™
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
EcoSafPak Glass Bottle
Glass Bottle
Plastic drum
Specifications
Specifications
Boiling Point | 270.0°C |
Melting Point | 6.0°C |
Density | 1.0670g/mL |
Quantity | 1 L |
Flash Point | 274°C |
Packaging | EcoSafPak Glass Bottle |
Refractive Index | 1.4900 to 1.4920 |
Specific Gravity | 1.067 |
Viscosity | 240 mPa.s (25°C) |
UN Number | 3082 |
Show More |
Safety and Handling
Product Identifier
- Triton X-100
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'