missing translation for 'onlineSavingsMsg'
Learn More

Triton™ X-100, Thermo Scientific™

Product Code. 10541754
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
EcoSafPak Glass Bottle
Glass Bottle
Plastic drum
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Polyoxyethylene(10) octylphenyl ether
PubChem CID 5590
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Boiling Point 270.0°C
Melting Point 6.0°C
Density 1.0670g/mL
Quantity 2.5 L
Flash Point 274°C
Packaging Glass Bottle
Refractive Index 1.4900 to 1.4920
Specific Gravity 1.067
Viscosity 240 mPa.s (25°C)
Merck Index 15, 6850
Solubility Information Solubility in water: soluble. Other solubilities: soluble in toluene, xylene, trichloroethylene,, ethylene glycol, ethyl ether, ethyl alcohol,, isopropyl alcohol, ethylene dichloride and most, other solvnets., insoluble in kerosene, miniral spirits.
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
Formula Weight 646.85
Chemical Name or Material Triton™ X-100
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'