missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl)silane, 97%, Thermo Scientific Chemicals

Product Code. 11420627
Click to view available options
Quantity:
1 g
5 g
25 g

Tris(trimethylsilyl)silane is used in hydrosilylations of carbonyls, radical reactions, reductions of acid chlorides, and carbon-halogen bonds. It acts a mediator of radical reactions and considered to be a nontoxic substitute for tri-n-butylstannane in radical reactions. It forms complexes with transition metals and main group elements. Further, it is utilized in carbon-carbon bond forming reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(trimethylsilyl)silane is used in hydrosilylations of carbonyls, radical reactions, reductions of acid chlorides, and carbon-halogen bonds. It acts a mediator of radical reactions and considered to be a nontoxic substitute for tri-n-butylstannane in radical reactions. It forms complexes with transition metals and main group elements. Further, it is utilized in carbon-carbon bond forming reactions.

Solubility
Miscible with pentane, ether, toluene and tetrahydrofuran. Sparingly miscible with acetone and acetonitrile. Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1873-77-4
Molecular Formula C9H28Si4
Molecular Weight (g/mol) 248.66
MDL Number MFCD00077893
InChI Key SQMFULTZZQBFBM-UHFFFAOYSA-N
Synonym tris trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane, tris trimethylsilyl silicon, ttmss, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl-trisilane, hypersilyl, tris trimethylsilyl silyl radical, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane #, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilan-2-yl
IUPAC Name 1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane
SMILES C[Si](C)(C)[SiH]([Si](C)(C)C)[Si](C)(C)C

Specifications

Boiling Point 82°C to 84°C (12mmHg)
Quantity 25 g
UN Number UN1993
Beilstein 1923953
Flash Point 55°C (131°F)
Merck Index 14,9757
Solubility Information Miscible with pentane,ether,toluene and tetrahydrofuran. Sparingly miscible with acetone and acetonitrile. Immiscible with water.
Formula Weight 248.67
Percent Purity 97%
Refractive Index 1.49
Sensitivity Air Sensitive
Density 0.806
Chemical Name or Material Tris(trimethylsilyl)silane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'