missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsiloxy)silane, 97%, Thermo Scientific Chemicals

Product Code. 11309335
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1873-89-8
Molecular Formula C9H27O3Si4
Molecular Weight (g/mol) 295.65
MDL Number MFCD00054865
InChI Key XAASNKQYFKTYTR-UHFFFAOYSA-N
Synonym tris trimethylsiloxy silane, tris trimethylsilyloxy silane, tris trimethylsilyloxy silicon, 1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy trisiloxane, 1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy trisiloxane, tris trimethylsilyloxy silyl radical, trisiloxane, 1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy
PubChem CID 6365044
SMILES C[Si](C)(C)O[Si](O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Boiling Point 64°C (10mmHg)
Quantity 25 g
UN Number UN1993
Flash Point 50°C (122°F)
IUPAC Name tris(trimethylsilyloxy)silicon
Formula Weight 296.66
Percent Purity 97%
Refractive Index 1.3865
Density 0.852
Chemical Name or Material Tris(trimethylsiloxy)silane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'