missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIS Hydrochloride, Made from Tromethamine USP, Biotech Reagent Grade, J.T. Baker™
Produced under GMP guidelines, but for which there is no USP/NF compendia for the product
408.00 € - 1155.00 €
Chemical Identifiers
CAS | 1185-53-1 |
---|---|
Molecular Formula | C4H12ClNO3 |
Molecular Weight (g/mol) | 157.594 |
InChI Key | QKNYBSVHEMOAJP-UHFFFAOYSA-N |
Synonym | tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride |
PubChem CID | 93573 |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
SMILES | C(C(CO)(CO)N)O.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15548254
|
Avantor J.T.Baker
4106-01 |
500 g | Poly bottle |
408.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15558254
|
Avantor J.T.Baker
4106-05 |
2.5 kg | Poly Bottle |
1155.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
1185-53-1 | |
157.594 | |
tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride | |
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
C4H12ClNO3 | |
QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
93573 | |
C(C(CO)(CO)N)O.Cl |
Specifications
1185-53-1 | |
C4H12ClNO3 | |
tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride | |
C(C(CO)(CO)N)O.Cl | |
157.594 | |
157.6 | |
TRIS Hydrochloride, Made from Tromethamine USP |
147°C to 153°C | |
1L = 0.00kg | |
QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride | |
93573 | |
GMP manufactured product |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Tris Hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Tris hydrochloride