missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris-HCl, 1M Solution, pH 8.0, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
33.30 € - 70.50 €
Chemical Identifiers
CAS | 1185-53-1 |
---|---|
Molecular Formula | C4H12ClNO3 |
Molecular Weight (g/mol) | 157.594 |
InChI Key | QKNYBSVHEMOAJP-UHFFFAOYSA-N |
PubChem CID | 93573 |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
SMILES | C(C(CO)(CO)N)O.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15893661
|
Thermo Scientific
J22638.AE |
100 mL | Nalgene bottle |
33.30 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15835398
|
Thermo Scientific
J22638.AP |
500 mL | Nalgene bottle |
56.70 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15855478
|
Thermo Scientific
J22638.K2 |
1 L | Nalgene bottle |
70.50 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
1185-53-1 | |
157.594 | |
93573 | |
C(C(CO)(CO)N)O.Cl |
C4H12ClNO3 | |
QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
Specifications
1185-53-1 | |
8 | |
C4H12ClNO3 | |
C(C(CO)(CO)N)O.Cl | |
157.594 | |
157.6 | |
Tris-HCl |
1 M | |
Solution | |
QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride | |
93573 | |
Molecular Biology |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only