missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III), 98%, Thermo Scientific Chemicals

Product Code. 11317928
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14319-13-2
Molecular Formula C33H60LaO6
Molecular Weight (g/mol) 691.742
MDL Number MFCD00010466
InChI Key VTNJXVDFRGBYJC-LWTKGLMZSA-N
Synonym Lanthanum(III)-DPM; La(TMHD)3
PubChem CID 72376397
IUPAC Name (Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;lanthanum
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[La]

Specifications

Melting Point 240°C
Boiling Point 370°C (decomposition)
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 688.72
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'